Would you like to log in again?
Float Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
456398 456395
76169-974PK 29.33 USD InStock
76169-974 76169-972
Float Racks for Microtubes, Globe Scientific
Racks Tube Racks
  • Autoclavable polypropylene construction for strength, buoyancy, and temperature resistance
  • Racks float with full load of filled tubes
  • Molded handle in center of rack for comfort and support during transport
  • Legs allow rack to stand upright for use on benchtop
  • Use in ice, water baths, or on the benchtop
  • 65 mm round rack fits most 400 mL beakers
  • Commonly found in cloning, microbiology, and immunology labs
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty