Would you like to log in again?
Place Microscope Slide Files, 20, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
513029A 513029B 513029W 513029Y 513029G
76169-998CS 216.17 USD InStock
76169-998 76170-000 76170-002 76170-004 76169-996
Place Microscope Slide Files, 20, Globe Scientific
Slide Holders
The durable 20-Place microscope slide file provides added protection to slides during routine storage.
  • Holds up to 20 microscope slides horizontally
  • Each space is pre-numbered for easy identification
  • Transparent cover protects slides and allows for easy reading of ID labels
  • Covers can be swung behind the base for ready access to the slides
  • Five color options for organization and classification of slides
The tray is produced from polystyrene (PS) and the lid is produced from styrene-acrylonitrile (SAN) which provides a longer usable life.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty