Would you like to log in again?
Heavy Duty Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
456320B 456320R 456320W
76170-052EA 28.53 USD InStock
76170-052 76170-054 76170-056
Heavy Duty Racks for Microtubes, Globe Scientific
Racks Tube Racks
This popular, rugged, reinforced polypropylene rack provides strength and durability.
  • Autoclavable
  • Rubber feet provide stability on slick surfaces
  • Numeric markings for sample identification
  • Compatible with both 1.5 and 2.0 ml microtubes
With rubber feet providing stability, these racks can be used as a workstation on the benchtop.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty