Would you like to log in again?
TUFFBLOK™ 4-Way Interlocking Tube Racks, Globe Scientific
Supplier: Globe Scientific
456940P 456940B 456940N 456940G 456940Y 456940AST 456940C
76170-120PK 64.49 USD InStock
76170-120 76170-122 76170-124 76170-114 76170-126 76170-116 76170-118
TUFFBLOK™ 4-Way Interlocking Tube Racks, Globe Scientific
Racks Tube Racks
These super strong and versatile TUFFBLOK™ 4-way tube racks hold variety of tube sizes in a single rack. Just flip the rack to accommodate the tubes you're using, and get to work!
  • Link multiple racks together for a custom rack solution
  • Produced from autoclavable polypropylene
  • Heavy-duty construction is durable and chemical resistant
  • Multiple colors available for benchtop organization at a glance
  • Numeric markings for sample identification
The 4 sides hold the following tubes: 4×50 ml tubes, 12×15 ml tubes, 32×1.5 ml tubes, and 32×0.5 ml tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty