Would you like to log in again?
Stands for Test Tubes, Globe Scientific
Supplier: Globe Scientific
456120B 456116R 456116B 456132B 456113R 456132W 456113W 456125Y 456116W 456113B 456120R 456125W 456132R 456125R 456132Y 456116Y 456113Y 456125B 456120W 456120Y
76170-156EA 17.35 USD InStock
76170-156 76170-178 76170-168 76170-158 76170-170 76170-182 76170-160 76170-172 76170-162 76170-184 76170-262 76170-174 76170-164 76170-186 76170-154 76170-264 76170-176 76170-166 76170-188 76170-180
Stands for Test Tubes, Globe Scientific
Racks Tube Racks
Separate and organize your tube samples by size with the help of these popular tube stands.
  • Three tier design holds tubes securely
  • Sturdy handles facilitate easy transport
  • Four color options for organization of samples
These rugged and versatile tube stands are produced from reinforced polypropylene and are fully autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty