Would you like to log in again?
Bard-Parker® Blade Remover, Aspen Surgical
Supplier: Aspen Surgical
Bard-Parker®
ST-115 ST-115NS
76170-430BX 420.1 USD InStock
76170-430 76170-278
Bard-Parker® Blade Remover, Aspen Surgical
Blade Removers and Dispensers
Helps safely remove up to five blades from conventional scalpels without the need to touch the blade.
  • Clear lid allows easy counting of blades that have been removed
  • Locking tabs prevent blades from being removed from device and clearly indicate when device is full
  • Adhesive backing secures device to any surface, including drapes
  • Features an audible and tactile response to ensure that blade is safely secure in device before removing scalpel handle
  • Intended for single-use and not made with natural rubber latex
Designed to safely unload contaminated blades into a secure channel, helping protect users from sharps injuries.
Helps to comply with OSHA Bloodbourne Standard 1910.1030(d)(2)(vii)(B) and AORN Recommended Practices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty