Would you like to log in again?
Citrullinated Histone H3 (Clone 11D3) ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
501620-96
76170-290EA 746.17 USD InStock
76170-290
Citrullinated Histone H3 (Clone 11D3) ELISA Kit, Cayman Chemical Company
Assays ELISAs
An ELISA for quantifying Citrullinated Histone H3 in biological matrices.
  • Detect CitH3 in cell culture supernatants and cell lysates
  • Detect CitH3 from as few as 1,000 neutrophils in human and mouse
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure CitH3 levels down to 0.15 ng/mL
  • Rapid assay; get results in under 4 hours
Histone H3 is citrullinated by PAD4 at residues R2, R8, and R17 during the processes of epigenetic chromatin remodeling and NET formation. In vitro, CitH3 is released from neutrophils that have been stimulated with calcium ionophore during the process of NETosis, and can be freed from NETs by treatment with nuclease. The measurement of CitH3, either in cell lysates or released during NETosis can be used as a functional readout of pharmacological PAD inhibition.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty