Would you like to log in again?
Phagocytosis Assay Kit (IgG FITC), Cayman Chemical Company
Supplier: Cayman Chemical
500290-1
76170-292EA 389.24 USD InStock
76170-292
Phagocytosis Assay Kit (IgG FITC), Cayman Chemical Company
Assays Cellular Assays
For the identification of factors regulating the phagocytic process in vitro.
  • Detect phagocytosis of fluorescein-labeled rabbit IgG-coated latex beads​
  • Validated in human peripheral blood monocytes and mouse peritoneal macrophages
  • Phagocytosis detectable in 1 – 4 hours at 37 °C
  • Assess by flow cytometry or fluorescence microscopy
Cayman’s Phagocytosis Assay Kit (IgG FITC) employs latex beads coated with fluorescently-labeled rabbit IgG as a probe for the measurement of the phagocytic process in vitro. The engulfed fluorescent beads can be detected using a fluorescence microscope, allowing kinetic studies of phagocytosis at the single-cell level. In addition, the flow cytometric readout provides the advantage of visualizing perturbations in phagocytosis on the population level and, when combined with antibody staining, of specific cell types within complex populations. This kit provides enough Latex Beads-Rabbit IgG-FITC Complex for up to 750 samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty