Clear, flint glass French square bottles provide maximum content visibility.
- Space saving design saves on shelf and storage space
The wide mouth opening is ideal for mixing, storing, and sampling.
Ordering information: Bottles 16152-188 and 16152-192 are supplied Ultra Cleaned. Bottles 16151-410, 36319-270, 36319-279, 36319-281, 36319-285, and 36319-288 are supplied pre-cleaned for use with volatiles.