Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JAK1 IgG Polyclonal Antibody

Supplier: Boster
A00330
76170-966EA 622.93 USD InStock
76170-966
Anti-JAK1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Tyrosine-protein kinase JAK1(JAK1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: JAK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty