Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig porcine TGF Beta 1 PicoKine; ELISA Kit, Boster Biological Technology

Supplier: Boster
PicoKine
EK0513-PO
76172-148EA 729.69 USD InStock
76172-148
Pig porcine TGF Beta 1 PicoKine; ELISA Kit, Boster Biological Technology
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of activated Pig porcine TGF beta 1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Pig Tgf Beta 1/TGFB1 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tgf Beta 1/TGFB1 tissue specificity: Highly expressed in bone. Abundantly expressed in articular cartilage and chondrocytes and is increased in osteoarthritis (OA). Colocalizes with ASPN in chondrocytes within OA lesions of articular cartilage. .

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty