Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse SPINT1/HAI-1 PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK0772
76172-222EA 729.72 USD InStock
76172-222
Mouse SPINT1/HAI-1 PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse SPINT1/HAI-1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Hai-1/Spint1 with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Hai-1/Spint1 tissue specificity:

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty