Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Enhanced Chemiluminescent Reagent Kit (Mouse IgG), Boster Biological Technology

Supplier: Boster
EK1001
76172-304EA 197.75 USD InStock
76172-304
Enhanced Chemiluminescent Reagent Kit (Mouse IgG), Boster Biological Technology
Assays ELISAs
Enhanced Chemiluminescent Reagent Kit (Mouse IgG)

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Enhanced Chemiluminescent Reagent Kit (Mouse IgG) for Western Blotting, with the specific secondary antibody and all the reagents needed for performing WB. 96wells/kit, with removable strips.

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty