Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse ALCAM/Cd166 PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK1184
76172-398EA 729.49 USD InStock
76172-398
Mouse ALCAM/Cd166 PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse ALCAM

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Cd166/Alcam with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cd166/Alcam tissue specificity: Detected on brain motor neurons, in differentiating retinal ganglion cells and in adult retina (PubMed:15345243). Detected on leukocytes and on lymphatic endothelial cells (PubMed:23169771). Detected in spleen B cells and T-cells (at protein level) (PubMed:9209500). Detected in adult brain and embryonic spinal cord (PubMed:15345243). Expressed at high levels in the brain, and lung, and at lower levels in the liver, and the kidney, as well as by activated leukocytes (PubMed:9209500). .

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty