Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Meteorin/METRN PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK1551
76172-696EA 729.49 USD InStock
76172-696
Mouse Meteorin/METRN PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse Meteorin/METRN

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Meteorin/Metrn with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Meteorin/Metrn tissue specificity: Highly expressed in brain. Expressed in undifferentiated neural progenitors and in astrocyte lineage, particulary in Bergmann glia, a subtype of radial glia, and a few discrete neuronal populations residing in the superior colliculus, the ocular motor nucleus, the raphe and pontine nuclei, and in various thalamic nuclei. Weakly expressed in heart, kidney, skeletal muscle, spleen, testis, gut and lung. .

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty