Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human S100A8/Calgranulin A PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK1558
76172-706EA 729.72 USD InStock
76172-706
Human S100A8/Calgranulin A PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human S100A8

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Human Calgranulin A/S100A8 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Calgranulin A/S100A8 tissue specificity: Calprotectin (S100A8/9) is predominantly expressed in myeloid cells. Except for inflammatory conditions, the expression is restricted to a specific stage of myeloid differentiation since both proteins are expressed in circulating neutrophils and monocytes but are absent in normal tissue macrophages and lymphocytes. Under chronic inflammatory conditions, such as psoriasis and malignant disorders, also expressed in the epidermis. Found in high concentrations at local sites of inflammation or in the serum of patients with inflammatory diseases such as rheumatoid, cystic fibrosis, inflammatory bowel disease, Crohn's disease, giant cell arteritis, cystic fibrosis, Sjogren's syndrome, systemic lupus erythematosus, and progressive systemic sclerosis. Involved in the formation and deposition of amyloids in the aging prostate known as corpora amylacea inclusions. Strongly up-regulated in many tumors, including gastric, esophageal, colon, pancreatic, bladder, ovarian, thyroid, breast and skin cancers. .

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty