Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse TNFRSF19/TROY PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK1585
76172-746EA 729.72 USD InStock
76172-746
Mouse TNFRSF19/TROY PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse TNFRSF19/TROY

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Tnfrsf19 with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tnfrsf19 tissue specificity: Highly expressed in adult brain, and in embryos from day 11-17, but not earlier. Detected in embryonic brain and epithelium, and at lower levels in adult heart, lung and liver. In neonatal mice, mainly in hair follicles and neuron-like cells in the cerebellum, but not in the skin epidermis. Isoform 3 was found in embryonic day 17.5 skin but not in brain and liver.

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty