Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CXCL9/Mig EZ-Set; ELISA Kit (DIY Antibody Pairs), Boster

Supplier: Boster
EZ0732
76172-822EA 425.04 USD InStock
76172-822
Human CXCL9/Mig EZ-Set; ELISA Kit (DIY Antibody Pairs), Boster
Assays ELISAs
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects CXCL9 in Human samples

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects CXCL9 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.

The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty