Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDAH2 IgG Polyclonal Antibody

Supplier: Boster
PB10001
76172-884EA 698.31 USD InStock
76172-884
Anti-DDAH2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for N(G),N(G)-dimethylarginine dimethylaminohydrolase 2(DDAH2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

1. Leiper, J. M,, Santa Maria, J., Chubb, A., MacAllister, R. J., Charles, I. G., Whitley, G. S., Vallance, P. Identification of two human dimethylarginine dimethylaminohydrolases with distinct tissue distributions and homology with microbial arginine deiminases.Biochem. J. 343: 209-214, 1999.
2. Smith, C. L., Birdsey, G. M., Anthony, S., Arrigoni, F. I., Leiper, J. M., Vallance, P. Dimethylarginine dimethylaminohydrolase activity modulates ADMA levels, VEGF expression, and cell phenotype. Biochem. Biophys. Res. Commun. 308: 984-989, 2003.
3. Tran, C. T. L., Fox, M. F., Vallance, P., Leiper, J. M. Chromosomal localization, gene structure, and expression pattern of DDAH1: comparison with DDAH2 and implications for evolutionary origins. Genomics 68: 101-105, 2000.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: DDAH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty