Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMGB2 IgG Polyclonal Antibody

Supplier: Boster
PB10002
76172-886EA 698.31 USD InStock
76172-886
Anti-HMGB2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for High mobility group protein B2(HMGB2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

1. "Entrez Gene: HMGB2 high-mobility group box 2".
2. Majumdar A, Brown D, Kerby S, Rudzinski I, Polte T, Randhawa Z, Seidman MM (Dec 1991). "Sequence of human HMG2 cDNA". Nucleic Acids Research 19 (23): 6643.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: HMGB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty