Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-INA IgG Polyclonal Antibody

Supplier: Boster
PB10003
76172-888EA 698.31 USD InStock
76172-888
Anti-INA IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Alpha-internexin(INA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

1. Chan, S.-O., Chiu, F.-C. Cloning and developmental expression of human 66 kd neurofilament protein. Molec. Brain Res. 29: 177-184, 1995.
2. Levavasseur F, Zhu Q, and JP Julien. No requirement of alpha-internexin for nervous system development and for radial growth of axons. Molecular Brain Research. 69:104-112. (1999).

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: INA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty