Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPSAB1 IgG Polyclonal Antibody

Supplier: Boster
PB10016
76172-914EA 698.31 USD InStock
76172-914
Anti-TPSAB1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Tryptase alpha/beta-1(TPSAB1) detection. Tested with WB, IHC-P, ELISA in Human.

1. 'Entrez Gene: TPSAB1 tryptase alpha/beta 1'.
2. Miller JS, Moxley G, Schwartz LB (Oct 1990). 'Cloning and characterization of a second complementary DNA for human tryptase'. J Clin Invest 86 (3): 864–70.
3. Pallaoro M, Fejzo MS, Shayesteh L, Blount JL, Caughey GH (Feb 1999). 'Characterization of genes encoding known and novel human mast cell tryptases on chromosome 16p13.3'. J Biol Chem 274 (6): 3355–62.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: TPSAB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty