Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Fetuin A Polyclonal Antibody

Supplier: Boster
PB9242
76172-958EA 622.93 USD InStock
76172-958
Anti-Fetuin A Polyclonal Antibody
Antibodies
Polyclonal antibody for 2017 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. 2017 information: Molecular Weight: 37326 MW; Subcellular Localization: Secreted; Tissue Specificity: Liver is the major site of synthesis, but fetuin is also expressed in limb buds and other extrahepatic tissues during development.

Type: Primary
Antigen: Fetuin A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty