Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WNT2B IgG Polyclonal Antibody

Supplier: Boster
PB9462
76172-986EA 698.31 USD InStock
76172-986
Anti-WNT2B IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Protein Wnt-2b(WNT2B) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

WNT2B (Wingless-Type MMTV Integration Site Family Member 2B), is a protein that in humans is encoded by the WNT2B gene. This gene encodes a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. Katoh et al. (1996) used fluorescence in situ hybridization to map the WNT13 gene to chromosome 1p13. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. This gene may play a role in human development as well as human carcinogenesis.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.

WB: The detection limit for Wnt2b is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: XBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty