Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFkB p100/p52 Polyclonal Antibody

Supplier: Boster
PB9150
76173-180EA 698.31 USD InStock
76173-180
Anti-NFkB p100/p52 Polyclonal Antibody
Antibodies
Polyclonal antibody for NFKB P100/NFKB2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. NFKB P100/NFKB2 information: Molecular Weight: 33233 MW; Subcellular Localization: Cytoplasm ; Tissue Specificity: Ubiquitous.

Type: Primary
Antigen: NFkB p100/p52
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty