Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Haptoglobin Polyclonal Antibody

Supplier: Boster
PB9219
76173-304EA 698.31 USD InStock
76173-304
Anti-Haptoglobin Polyclonal Antibody
Antibodies
Polyclonal antibody for HAPTOGLOBIN/HP detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. HAPTOGLOBIN/HP information: Molecular Weight: 45205 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed by the liver and secreted in plasma.

Type: Primary
Antigen: Haptoglobin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty