Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IKK Beta Polyclonal Antibody

Supplier: Boster
PB9221
76173-308EA 698.31 USD InStock
76173-308
Anti-IKK Beta Polyclonal Antibody
Antibodies
Polyclonal antibody for IKK beta/IKBKB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. IKK beta/IKBKB information: Molecular Weight: 86564 MW; Subcellular Localization: Cytoplasm. Nucleus. Membrane raft. Colocalized with DPP4 in membrane rafts; Tissue Specificity: Highly expressed in heart, placenta, skeletal muscle, kidney, pancreas, spleen, thymus, prostate, testis and peripheral blood.

Type: Primary
Antigen: IKK Beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty