Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NR3C1 Polyclonal Antibody

Supplier: Boster
PB9232
76173-330EA 698.31 USD InStock
76173-330
Anti-NR3C1 Polyclonal Antibody
Antibodies
Polyclonal antibody for GLUCOCORTICOID RECEPTOR/NR3C1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GLUCOCORTICOID RECEPTOR/NR3C1 information: Molecular Weight: 85659 MW; Subcellular Localization: Cytoplasm . Mitochondrion. Nucleus . Cytoplasmic in the absence of ligand, nuclear after ligand- binding; Tissue Specificity: Widely expressed. In the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart.

Type: Primary
Antigen: NR3C1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty