Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRPC5 Polyclonal Antibody

Supplier: Boster
PB9271
76173-376EA 622.76 USD InStock
76173-376
Anti-TRPC5 Polyclonal Antibody
Antibodies
Polyclonal antibody for TRPC5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. TRPC5 information: Molecular Weight: 111412 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein ; Tissue Specificity: Expressed in brain with higher levels in fetal brain. Found in cerebellum and occipital pole.

Type: Primary
Antigen: TRPC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty