Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIM1 Polyclonal Antibody

Supplier: Boster
PB9315
76173-464EA 622.93 USD InStock
76173-464
Anti-PIM1 Polyclonal Antibody
Antibodies
Polyclonal antibody for PIM1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PIM1 information: Molecular Weight: 45412 MW; Subcellular Localization: Isoform 2: Cytoplasm. Nucleus; Tissue Specificity: Expressed primarily in cells of the hematopoietic and germline lineages. Isoform 1 and isoform 2 are both expressed in prostate cancer cell lines.

Type: Primary
Antigen: PIM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty