Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTCH1 IgG Polyclonal Antibody

Supplier: Boster
PB9352
76173-538EA 623.01 USD InStock
76173-538
Anti-PTCH1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Protein patched homolog 1(PTCH1) detection. Tested with WB in Human;Mouse;Rat.

PTCH1 is also known as PTC, BCNS or HPE7. This gene encodes a member of the patched gene family. The encoded protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis, as well as the desert hedgehog and indian hedgehog proteins. This gene functions as a tumor suppressor. Mutations of this gene have been associated with basal cell nevus syndrome, esophageal squamous cell carcinoma, trichoepitheliomas, transitional cell carcinomas of the bladder, as well as holoprosencephaly. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences and biological validity cannot be determined currently.

Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.

Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05 mg NaN3.

WB: The detection limit for Patched is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SEMA3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty