Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBP4 IgG Polyclonal Antibody

Supplier: Boster
PB9360
76173-554EA 622.92 USD InStock
76173-554
Anti-RBP4 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Retinol-binding protein 4(RBP4) detection. Tested with WB, ELISA in Human;Mouse;Rat.

RBP4 is also known as RDCCAS. This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.

WB: The detection limit for RBP4 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: PRL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Yes
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty