Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRDX5 Rabbit Polyclonal Antibody

Supplier: Boster
PB9384
76173-602EA 698.31 USD InStock
76173-602
Anti-PRDX5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Peroxiredoxin-5, mitochondrial(PRDX5) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.

PRDX5 (peroxiredoxin 5) also known as AOEB166, ACR1,B166, mgC117264, mgC142283, mgC142285, PLP, PMP20, PRDX6, PRXV, SBBI10, is a member of the peroxiredoxin family and may play an antioxidant protective role in various tissues under nonpathologic conditions and during inflammatory processes. The PRDX5 gene is mapped to11q13.1. PRDX5 displays mitochondrial presequence features and has 3 cysteines implicated in antioxidant activity and a C-terminal SQL peroxisomal targeting sequence. Northern blot analysis revealed ubiquitous expression of a 1.0-kb PRDX5 transcript in tissues and cell lines. Functional analysis showed that PRDX5 has antioxidant activity equivalent to that of CAT. While PRDX5 was localized to fibroblasts in normal tendon, it was localized to fibroblasts and endothelial cells in degenerative tendon. PRDX5 mRNA and protein levels increased at 12 hours, and the increase in PRDX5 expression correlated with reduced peroxide levels. PRDX5 plays a protective role against oxidative stress in human cartilage.

Add 0.2 ml of distilled water will yield a concentration of 500µg/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

WB: The detection limit for Peroxiredoxin 5 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
, Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty