Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANXA3 Rabbit Polyclonal Antibody

Supplier: Boster
PB9420
76173-672EA 698.31 USD InStock
76173-672
Anti-ANXA3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Annexin A3(ANXA3) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Annexin A3 is a protein that in humans is encoded by the Annexin A3 gene. The Annexin A3 gene contains 13 exons and spans 58 kb of genomic DNA. The Annexin A3 gene is mapped to 4q21. It is abnormally expressed in fetuses of both IVF and ICSI, which may contribute to the increase risk of birth defects in these ART. This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phospholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.

Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: APH1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty