Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATXN3 Rabbit Polyclonal Antibody

Supplier: Boster
PB9423
76173-678EA 698.31 USD InStock
76173-678
Anti-ATXN3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Ataxin-3(ATXN3) detection. Tested with WB, IHC-P, IHC-F, ICC, FCM in Human;Rat.

ATXN3 (Ataxin 3), also known as AT3, MJD GENE, MJD1, SCA3 GENE, ATX3, JOS, Spinocerebellar ataxia-3, Machado-Joseph disease protein 1, is a protein that in humans is encoded by the ATXN3 gene. ATXN3 ranges in size from 360 to 374 amino acids. Using Northern blot analysis showed that ATXN3 mRNA was ubiquitously expressed in human tissues. They detected at least 4 ATXN3 transcripts of 1.4, 1.8, 4.5, and 7.5 kb and suggested that the different mRNA species probably result from differential splicing and polyadenylation. Machado-Joseph disease, also known as spinocerebellar ataxia-3, is an autosomal dominant neurologic disorder. The protein encoded by the ATXN3 gene contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is an inverse correlation between the age of onset and CAG repeat numbers. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. Ataxin-3 interacted with 2 human homologs of the yeast DNA repair protein RAD23, HHR23A (RAD23A) and HHR23B (RAD23B). Both normal and mutant ataxin-3 proteins interacted with the ubiquitin-like domain at the N terminus of the HHR23 proteins, which is a motif important for nucleotide excision repair. However, in HEK 293 cells, HHR23A was recruited to intranuclear inclusions formed by the mutant ataxin-3 through its interaction with ataxin-3.

Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: CCNA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty