Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Lcat IgG Polyclonal Antibody

Supplier: Boster
PB9657
76173-778EA 622.93 USD InStock
76173-778
Anti-Lcat IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Phosphatidylcholine-sterol acyltransferase(LCAT) detection. Tested with WB in Human;Mouse;Rat.

LCAT (Lecithin: Cholesterol Acyltransferase), is an enzyme that converts free cholesterol into cholesteryl ester. Azoulay et al. (1987) used a cDNA clone corresponding to LCAT to assign the locus to 16q22 through the analysis of DNA from somatic cell hybrids and in situ hybridization. LCAT plays an important role in lipoprotein metabolism, especially in the process termed 'reverse cholesterol transport.' The enzyme is synthesized in the liver and circulates in blood plasma as a complex with components of high density lipoprotein (HDL). Cholesterol from peripheral cells is transferred to HDL particles, esterified through the action of LCAT on HDL, and incorporated into the core of the lipoprotein. The cholesterol ester is thereby transported to the liver (Jonas, 2000).

Type: Primary
Antigen: Lcat
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty