Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LYZ IgG Polyclonal Antibody

Supplier: Boster
PB9663
76173-790EA 698.31 USD InStock
76173-790
Anti-LYZ IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Lysozyme C(LYZ) detection. Tested with WB, IHC-P in Human;Rat.

In humans, the lysozyme enzyme is encoded by the LYZ gene. This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta [1-4] glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the antimicrobial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. The protein has antibacterial activity against a number of bacterial species. Missense mutations in this gene have been identified in heritable renal amyloidosis.

Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty