Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAOA IgG Polyclonal Antibody

Supplier: Boster
PB9664
76173-792EA 698.31 USD InStock
76173-792
Anti-MAOA IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Amine oxidase [flavin-containing] A(MAOA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

MAOA(Monoamine oxidase A), also known as AMINE OXIDASE (FLAVIN-CONTAINING) A, is an enzyme that in humans is encoded by the MAO-A gene. MAOA is an isozyme of monoamine oxidase which is also mapped on Xp11.3. MAOA degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the outer mitochondrial membrane. Mutation in MAOA results in monoamine oxidase deficiency, or Brunner syndrome. In humans, there is a 30-base repeat sequence repeated in one of several different numbers of times in the promoter region of the gene coding for MAOA. MAO-A levels in the brain as measured using positron emission tomography are elevated by an average of 34% in patients with major depressive disorder. Inhibition of MAOA prevented apoptosis, and serum starvation of cortical brain cells from Maoa-deficient mice resulted in reduced apoptosis compared with wildtype mice.

Type: Primary
Antigen: MAOA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty