Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NLRC4 IgG Polyclonal Antibody

Supplier: Boster
PB9673
76173-810EA 622.76 USD InStock
76173-810
Anti-NLRC4 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for NLR family CARD domain-containing protein 4(NLRC4) detection. Tested with WB in Human;Mouse;Rat.

NLR family CARD domain-containing protein 4, also known as CARD12, is a protein that in humans is encoded by the NLRC4 gene. It is mapped to 2p22.3. This gene encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.

Type: Primary
Antigen: NLRC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty