Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GREM1 IgG Polyclonal Antibody

Supplier: Boster
PB9626
76174-108EA 622.93 USD InStock
76174-108
Anti-GREM1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Gremlin-1(GREM1) detection. Tested with WB in Human;Rat.

Gremlin, also known as Drm, is a highly conserved 20.7-kDa, 184 amino acid glycoprotein part of the DAN family and is a cysteine knot-secreted protein. Skeletal cells synthesize bone morphogenetic proteins (BMPs) and BMP antagonists. And Gremlin is expressed in osteoblasts and opposes BMP effects on osteoblastic differentiation and function in vitro. Gremlin 1 (GREM 1) is known for its antagonistic reaction with BMPs in the TGF beta signaling pathway. This gene inhibits BMP-2, BMP-4, and BMP-7. Inhibition by grem 1 of BMPs in mice allow the expression of fibroblast growth factors (FGFs) 4 and 8 and Sonic hedgehog (SHH) which are necessary for proper limb development. Gremlin 1 may play an oncogenic role especially in carcinomas of the uterine cervix, lung, ovary, kidney, breast, colon, pancreas, and sarcoma. Over-expressed gremlin 1 functions by interaction with YWHAH (Its binding site for gremlin 1 was located between residues 61-80 and gremlin 1 binding site for YWHAH was found to be located between residues 1-67). Therefore, Gremlin 1 and its binding protein YWHAH could be good targets for developing diagnostic and therapeutic strategies against human cancers.

Type: Primary
Antigen: GREM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty