Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABR IgG Polyclonal Antibody

Supplier: Boster
PB9972
76174-406EA 698.31 USD InStock
76174-406
Anti-ABR IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Active breakpoint cluster region-related protein(ABR) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

1. Heisterkamp, N., Kaartinen, V., van Soest, S., Bokoch, G. M., Groffen, J. Human ABR encodes a protein with GAP-rac activity and homology to the DBL nucleotide exchange factor domain. J. Biol. Chem. 268: 16903-16906, 1993.
2. Heisterkamp, N., Morris, C., Groffen, J. ABR, an active BCR-related gene. Nucleic Acids Res. 17: 8821-8831, 1989.
3. McDonald, J. D., Daneshvar, L., Willert, J. R., Matsumura, K., Waldman, F., Cogen, P. H. Physical mapping of chromosome 17p13.3 in the region of a putative tumor suppressor gene important in medulloblastoma. Genomics 23: 229-232, 1994.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ABR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty