Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGO4 IgG Polyclonal Antibody

Supplier: Boster
PB9979
76174-420EA 622.93 USD InStock
76174-420
Anti-AGO4 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Protein argonaute-4(AGO4) detection. Tested with WB in Human;Rat.

1. Carmell, M. A., Xuan, Z., Zhang, M. Q., Hannon, G. J. The argonaute family: tentacles that reach into RNAi, developmental control, stem cell maintenance, and tumorigenesis. Genes Dev. 16: 2733-2742, 2002.
2. Nagase, T., Kikuno, R., Nakayama, M., Hirosawa, M., Ohara, O. Prediction of the coding sequences of unidentified human genes. XVIII. The complete sequences of 100 new cDNA clones from brain which code for large proteins in vitro. DNA Res. 7: 273-281, 2000.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: AGO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty