Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AIRE IgG Polyclonal Antibody

Supplier: Boster
PB9980
76174-422EA 698.31 USD InStock
76174-422
Anti-AIRE IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Autoimmune regulator(AIRE) detection. Tested with WB in Human;Rat.

1. Lindmark E, Chen Y, Georgoudaki AM, Dudziak D, Lindh E, Adams WC, Loré K, Winqvist O, Chambers BJ, Karlsson MC, et al. (2013). "AIRE expressing marginal zone dendritic cells balances adaptive immunity and T-follicular helper cell recruitment.". Journal of Autoimmunity 42: 62–70.
2. Liston A, Lesage S, Wilson J, Peltonen L, Goodnow CC (April 2003). "Aire regulates negative selection of organ-specific T cells". Nat. Immunol. 4(4): 350–4.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: AIRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty