Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCR IgG Polyclonal Antibody

Supplier: Boster
PB9988
76174-836EA 622.93 USD InStock
76174-836
Anti-BCR IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Breakpoint cluster region protein(BCR) detection. Tested with WB in Human;Mouse;Rat.

1. "Entrez Gene: Breakpoint cluster region".
2. Puil L, Liu J, Gish G, Mbamalu G, Bowtell D, Pelicci PG, Arlinghaus R, Pawson T (February 1994). "Bcr-Abl oncoproteins bind directly to activators of the Ras signalling pathway". EMBO J. 13 (4): 764–73.
3. Pendergast AM, Muller AJ, Havlik MH, Maru Y, Witte ON (July 1991). "BCR sequences essential for transformation by the BCR-ABL oncogene bind to the ABL SH2 regulatory domain in a non-phosphotyrosine-dependent manner".Cell 66 (1): 161–71.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty