Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CA2 IgG Polyclonal Antibody

Supplier: Boster
PB9989
76174-838EA 698.31 USD InStock
76174-838
Anti-CA2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Carbonic anhydrase 2(CA2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

1. Aramaki, S., Yoshida, I., Yoshino, M., Kondo, M., Sato, Y., Noda, K., Jo, R., Okue, A., Sai, N., Yamashita, F. Carbonic anhydrase II deficiency in three unrelated Japanese patients. J. Inherit. Metab. Dis. 16: 982-990, 1993.
2. Laitala, T., Vaananen, H. K. Inhibition of bone resorption in vitro by antisense RNA and DNA molecules targeted against carbonic anhydrase II or two subunits of vacuolar H(+)-ATPase. J. Clin. Invest. 93: 2311-2318, 1994.
3. Venta, P. J., Welty, R. J., Johnson, T. M., Sly, W. S., Tashian, R. E. Carbonic anhydrase II deficiency syndrome in a Belgian family is caused by a point mutation at an invariant histidine residue (107his-to-tyr): complete structure of the normal human CA II gene. Am. J. Hum. Genet. 49: 1082-1090, 1991.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty