Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SFRP4 Polyclonal Antibody

Supplier: Boster
RP1067
76174-926EA 718.26 USD InStock
76174-926
Anti-SFRP4 Polyclonal Antibody
Antibodies
Polyclonal antibody for sFRP-4/SFRP4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. sFRP-4/SFRP4 information: Molecular Weight: 39827 MW; Subcellular Localization: Secreted . Cytoplasmic in ovarian tumor cells; Tissue Specificity: Expressed in mesenchymal cells. Highly expressed in the stroma of proliferative endometrium. Expressed in cardiomyocytes. Shows moderate to strong expression in ovarian tumors with expression increasing as the tumor stage increases. In ovarian tumors, expression levels are inversely correlated with expression of CTNNB1 (at protein level).

Type: Primary
Antigen: SFRP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty