Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SP1 Polyclonal Antibody

Supplier: Boster
RP1072
76174-936EA 718.26 USD InStock
76174-936
Anti-SP1 Polyclonal Antibody
Antibodies
Polyclonal antibody for SP1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. SP1 information: Molecular Weight: 80693 MW; Subcellular Localization: Nucleus. Cytoplasm. Nuclear location is governed by glycosylated/phosphorylated states. Insulin promotes nuclear location, while glucagon favors cytoplasmic location; Tissue Specificity: Up-regulated in adenocarcinomas of the stomach (at protein level). Isoform 3 is ubiquitously expressed at low levels.

Type: Primary
Antigen: SP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty