Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSG101 Polyclonal Antibody

Supplier: Boster
RP1078
76174-946EA 718.26 USD InStock
76174-946
Anti-TSG101 Polyclonal Antibody
Antibodies
Polyclonal antibody for TSG 101/TSG101 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. TSG 101/TSG101 information: Molecular Weight: 43944 MW; Subcellular Localization: Cytoplasm. Membrane; Peripheral membrane protein. Nucleus. Late endosome membrane; Peripheral membrane protein. Mainly cytoplasmic. Membrane-associated when active and soluble when inactive. Depending on the stage of the cell cycle, detected in the nucleus. Colocalized with CEP55 in the midbody during cytokinesis; Tissue Specificity: Heart, brain, placenta, lung, liver, skeletal, kidney and pancreas.

Type: Primary
Antigen: TSG101
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty