Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTOV1 Polyclonal Antibody

Supplier: Boster
RP1087
76174-962EA 718.26 USD InStock
76174-962
Anti-PTOV1 Polyclonal Antibody
Antibodies
Polyclonal antibody for PTOV1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PTOV1 information: Molecular Weight: 46869 MW; Subcellular Localization: Cell membrane. Cytoplasm, perinuclear region. Nucleus. Translocates from the cytoplasm to the nucleus at the onset of S-phase. Also localizes to lipid rafts; Tissue Specificity: Expressed in brain, heart, kidney, liver, placenta, skeletal muscle and small intestine.

Type: Primary
Antigen: PTOV1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty