Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ticam1 IgG Polyclonal Antibody

Supplier: Boster
RP1100
76174-986EA 718.26 USD InStock
76174-986
Anti-Ticam1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for TIR domain-containing adapter molecule 1(TICAM1) detection. Tested with WB in Mouse.

Type: Primary
Antigen: Ticam1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty